5-Chloro-2,3-dimethoxypyridine-4-boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (5-Chloro-2,3-dimethoxypyridin-4-yl)boronic acid

23332-1G 23332-25G 23332-5G
76928-012EA 102.91 USD
76928-012 76928-014 76928-016
5-Chloro-2,3-dimethoxypyridine-4-boronic acid ≥95%
5-Chloro-2,3-dimethoxypyridine-4-boronic acid

Precaución: for Research Use Only

Formula: C₇H₉BClNO₄
MW: 217.415 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20231439
CAS Number: 2096339-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR