4-Methoxy-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

23230-25G 23230-5G
76927-960EA 453.01 USD
76927-960 76927-962
4-Methoxy-2-methylphenol ≥97%
4-Methoxy-2-methylphenol
Formula: C₈H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD11055138
CAS Number: 5307-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR