5-Chloro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

23192-25G 23192-5G
76927-950EA 289.63 USD
76927-950 76927-952
5-Chloro-1H-indazole ≥97%
5-Chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
Storage Temperature: Ambient
MDL Number: MFCD00022785
CAS Number: 698-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR