4-Bromo-5-fluoro-2-methoxyphenol ≥97% research grade

Supplier: AOBChem USA

Warning

23000-25G 23000-5G
76927-916EA 346.92 USD
76927-916 76927-918
4-Bromo-5-fluoro-2-methoxyphenol ≥97% research grade
4-Bromo-5-fluoro-2-methoxyphenol
Formula: C₇H₆BrFO₂
MW: 221.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437599
CAS Number: 886510-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR