4-Fluoro-3,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

22814-25G 22814-5G
76927-892EA 516.66 USD
76927-892 76927-894
4-Fluoro-3,5-dimethylbenzoic acid ≥97%
4-Fluoro-3,5-dimethylbenzoic acid
Formula: C₉H₉FO₂
MW: 168.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00184969
CAS Number: 120512-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR