5-Bromo-4-methylpyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-4-methyl-3-pyridinol

22801-1G 22801-25G 22801-500MG 22801-5G
76927-880EA 350.1 USD
76927-880 76927-882 76927-884 76927-886
5-Bromo-4-methylpyridin-3-ol ≥97%
5-Bromo-4-methylpyridin-3-ol
Formula: C₆H₆BrNO
MW: 188.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18257654
CAS Number: 351458-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR