4-Fluoro-3-methoxy-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

22768-1G 22768-25G 22768-500MG 22768-5G
76927-860EA 290.69 USD
76927-860 76927-862 76927-864 76927-866
4-Fluoro-3-methoxy-5-methylphenylboronic acid ≥97%
4-Fluoro-3-methoxy-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD20526393
CAS Number: 1451392-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR