5-Bromo-2-fuoro-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

22766-1G 22766-25G 22766-5G
76927-848EA 70.02 USD
76927-848 76927-850 76927-852
5-Bromo-2-fuoro-3-methylphenol ≥97%
5-Bromo-2-fuoro-3-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.024 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20526391
CAS Number: 1351668-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR