1-Bromo-3-fluoro-2,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-fluoro-1,3-dimethoxybenzene

22753-1G 22753-25G 22753-500MG 22753-5G
76927-832EA 335.25 USD
76927-832 76927-834 76927-836 76927-838
1-Bromo-3-fluoro-2,4-dimethoxybenzene ≥97%
1-Bromo-3-fluoro-2,4-dimethoxybenzene
Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22056749
CAS Number: 222547-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR