5-Bromo-2,3-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22742-25G 22742-5G
76927-820EA 293.87 USD
76927-820 76927-822
5-Bromo-2,3-difluorobenzaldehyde ≥97%
5-Bromo-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MW: 220.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669446
CAS Number: 633327-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR