4-Bromo-7-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

22528-1G 22528-25G 22528-500MG 22528-5G
76927-746EA 260.99 USD
76927-746 76927-748 76927-750 76927-752
4-Bromo-7-methyl-1H-indazole ≥97%
4-Bromo-7-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD12028617
CAS Number: 1159511-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR