4-Ethoxy-3,5-difluorobromobenzene ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-ethoxy-1,3-difluorobenzene

22477-1G 22477-25G 22477-5G
76927-720EA 105.03 USD
76927-720 76927-722 76927-724
4-Ethoxy-3,5-difluorobromobenzene ≥95% research grade
4-Ethoxy-3,5-difluorobromobenzene
Formula: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18917873
CAS Number: 115467-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR