5-Bromo-1,7-dimethyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-1,7-dimethylindazole

22441-1G 22441-25G 22441-500MG 22441-5G
76927-694EA 144.29 USD
76927-694 76927-696 76927-698 76927-700
5-Bromo-1,7-dimethyl-1H-indazole ≥97%
5-Bromo-1,7-dimethyl-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₉BrN₂
MW: 225.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11226595
CAS Number: 1092352-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR