3-Amino-4-fluoro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-1H-indazol-3-amine

22191-25G 22191-5G
76927-628EA 516.66 USD
76927-628 76927-630
3-Amino-4-fluoro-1H-indazole ≥97%
3-Amino-4-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆FN₃
MW: 151.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00119934
CAS Number: 404827-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR