5-Chloro-4-hydroxy-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

22149-1G 22149-25G 22149-500MG 22149-5G
76927-612EA 165.51 USD
76927-612 76927-614 76927-616 76927-618
5-Chloro-4-hydroxy-2-methoxypyridine ≥97%
5-Chloro-4-hydroxy-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂
MW: 159.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189878
CAS Number: 1196146-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR