4-Iododibenzothiophene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Iododibenzo[b,d]thiophene

22077-25G 22077-5G
76927-602EA 247.19 USD
76927-602 76927-604
4-Iododibenzothiophene ≥97%
4-Iododibenzothiophene
Formula: C₁₂H₇IS
MW: 310.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159895
CAS Number: 132034-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR