4-Bromo-2-ethylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

22074-1G 22074-25G 22074-500MG 22074-5G
76927-588EA 231.28 USD
76927-588 76927-590 76927-592 76927-594
4-Bromo-2-ethylbenzenemethanol ≥97%
4-Bromo-2-ethylbenzenemethanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19105405
CAS Number: 877131-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR