4-Ethoxy-4'-hydroxybiphenyl ≥95%

Supplier: AOBChem USA

Warning

22027-1G 22027-25G 22027-500MG 22027-5G
76927-572EA 292.81 USD
76927-572 76927-574 76927-576 76927-578
4-Ethoxy-4'-hydroxybiphenyl ≥95%
4-Ethoxy-4'-hydroxybiphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200814
CAS Number: 127972-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR