5-Bromo-6-methylpicolinonitrile ≥97%

Supplier: AOBChem USA

Warning

21990-25G 21990-5G
76927-560EA 275.84 USD
76927-560 76927-562
5-Bromo-6-methylpicolinonitrile ≥97%
5-Bromo-6-methylpicolinonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD13181587
CAS Number: 1173897-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR