4-Bromo-4'-ethoxybiphenyl ≥97%

Supplier: AOBChem USA

Warning

21873-25G 21873-5G
76927-530EA 183.54 USD
76927-530 76927-532
4-Bromo-4'-ethoxybiphenyl ≥97%
4-Bromo-4'-ethoxybiphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD14582781
CAS Number: 58743-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR