5-Bromo-2,3,4-trifluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-2,3,4-trifluorophenyl)boronic acid, 5-Bromo-2,3,4-trifluorophenylboronic acid

21859-1G 21859-25G 21859-500MG 21859-5G
76927-514EA 296 USD
76927-514 76927-516 76927-518 76927-520
5-Bromo-2,3,4-trifluorobenzeneboronic acid ≥97%
5-Bromo-2,3,4-trifluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₃BBrF₃O₂
MW: 254.797 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12032026
CAS Number: 2377611-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR