2-Chloro-5-(hydroxymethyl)phenol ≥95%

Supplier: AOBChem USA

Warning

21602-1G 21602-25G 21602-5G
76927-470EA 88.06 USD
76927-470 76927-472 76927-474
2-Chloro-5-(hydroxymethyl)phenol ≥95%
2-Chloro-5-(hydroxymethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD18393156
CAS Number: 1261598-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR