5-Chloro-2-fluoro-3-(hydroxymethyl)pyridine ≥95% research grade

Supplier: AOBChem USA

Warning

21462-1G 21462-25G 21462-500MG 21462-5G
76927-442EA 171.87 USD
76927-442 76927-444 76927-446 76927-448
5-Chloro-2-fluoro-3-(hydroxymethyl)pyridine ≥95% research grade
5-Chloro-2-fluoro-3-(hydroxymethyl)pyridine
Formula: C₆H₅ClFNO
MW: 161.561 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972393
CAS Number: 884494-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR