4-Bromo-5-chloro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

21391-1G 21391-25G 21391-500MG 21391-5G
76927-426EA 183.54 USD
76927-426 76927-428 76927-430 76927-432
4-Bromo-5-chloro-1H-indazole ≥97%
4-Bromo-5-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12828060
CAS Number: 1056264-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR