(4-Butoxy-3-chlorophenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Butoxy-3-chlorophenylboronic acid

21217-25G 21217-5G
76927-402EA 240.83 USD
76927-402 76927-404
(4-Butoxy-3-chlorophenyl)boronic acid ≥97% research grade
(4-Butoxy-3-chlorophenyl)boronic acid
Formula: C₁₀H₁₄BClO₃
Storage Temperature: Ambient
MDL Number: MFCD04974064
CAS Number: 480438-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR