4-Carbamoyl-3-fluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Carbamoyl-3-fluorophenylboronic acid, (4-Carbamoyl-3-fluorophenyl)boronic acid

21202-1G 21202-25G 21202-500MG 21202-5G
76927-388EA 130.5 USD
76927-388 76927-390 76927-392 76927-394
4-Carbamoyl-3-fluorobenzeneboronic acid ≥97%
4-Carbamoyl-3-fluorobenzeneboronic acid
Formula: C₇H₇BFNO₃
MW: 182.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09258667
CAS Number: 874288-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR