4-Chloro-2,5-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

21165-1G 21165-25G 21165-500MG 21165-5G
76927-376EA 201.58 USD
76927-376 76927-378 76927-380 76927-382
4-Chloro-2,5-difluorophenylboronic acid ≥97%
4-Chloro-2,5-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BClF₂O₂
MW: 192.356 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18415647
CAS Number: 2055778-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR