5-(Methylsulfonyl)pyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

21145-1G 21145-25G 21145-500MG 21145-5G
76927-356EA 281.14 USD
76927-356 76927-358 76927-360 76927-362
5-(Methylsulfonyl)pyridine-3-boronic acid ≥97%
5-(Methylsulfonyl)pyridine-3-boronic acid
Formula: C₆H₈BNO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD08689536
CAS Number: 913836-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR