(5-Fluoro-1H-indazol-6-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

21115-1G 21115-500MG 21115-5G
76927-344EA 551.67 USD
76927-344 76927-346 76927-348
(5-Fluoro-1H-indazol-6-yl)boronic acid ≥95%
Formula: C₇H₆BFN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18089821
CAS Number: 1253911-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR