4-Butoxy-2,3-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Butoxy-2,3-difluorophenyl)boronic acid, 4-Butoxy-2,3-difluorobenzeneboronic acid

20889-25G 20889-5G
76927-270EA 346.92 USD
76927-270 76927-272
4-Butoxy-2,3-difluorophenylboronic acid ≥97%
4-Butoxy-2,3-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD19441186
CAS Number: 156487-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR