4-Ethoxy-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

20793-1G 20793-25G 20793-500MG 20793-5G
76927-242EA 159.14 USD
76927-242 76927-244 76927-246 76927-248
4-Ethoxy-3-fluorobenzaldehyde ≥97%
4-Ethoxy-3-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06247348
CAS Number: 452-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR