5-Chloro-2-fluoro-3-hydroxypyridine ≥97% research grade

Supplier: AOBChem USA

Warning

20727-1G 20727-25G 20727-5G
76927-228EA 117.76 USD
76927-228 76927-230 76927-232
5-Chloro-2-fluoro-3-hydroxypyridine ≥97% research grade
5-Chloro-2-fluoro-3-hydroxypyridine
Formula: C₅H₃ClFNO
Storage Temperature: Refrigerator
MDL Number: MFCD09835255
CAS Number: 1003711-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR