4-Bromo-3-(trifluoromethyl)benzoic acid ethyl ester ≥97%

Supplier: AOBChem USA

Warning

20702-25G 20702-5G
76927-216EA 524.09 USD
76927-216 76927-218
4-Bromo-3-(trifluoromethyl)benzoic acid ethyl ester ≥97%
4-Bromo-3-(trifluoromethyl)benzoic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09745861
CAS Number: 463355-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR