4-Fluoro-3-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

20632-1G 20632-25G 20632-500MG 20632-5G
76927-206EA 228.1 USD
76927-206 76927-208 76927-210 76927-212
4-Fluoro-3-methyl-1H-indazole ≥97%
4-Fluoro-3-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇FN₂
MW: 150.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04109834
CAS Number: 662146-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR