Methyl 5-bromo-2-(trifluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-(trifluoromethoxy)benzoic acid methyl ester

20618-1G 20618-25G 20618-500MG 20618-5G
76927-194EA 180.36 USD
76927-194 76927-196 76927-198 76927-200
Methyl 5-bromo-2-(trifluoromethoxy)benzoate ≥97%
Methyl 5-bromo-2-(trifluoromethoxy)benzoate
Formula: C₉H₆BrF₃O₃
MW: 299.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203799
CAS Number: 773874-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR