5-(tert-Butylcarbamoyl)-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(Tert-butylcarbmoyl)-2-fluorophenylboronic acid, 5-tert-Butylcarbamoyl-2-fluorobenzeneboronic acid, (5-(tert-Butylcarbamoyl)-2-fluorophenyl)boronic acid

20596-1G 20596-25G 20596-500MG 20596-5G
76927-178EA 142.17 USD
76927-178 76927-180 76927-182 76927-184
5-(tert-Butylcarbamoyl)-2-fluorophenylboronic acid ≥97%
5-(tert-Butylcarbamoyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₃
MW: 239.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436054
CAS Number: 874289-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR