5-Hydroxy-2-methoxy-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

20566-1G 20566-25G 20566-500MG 20566-5G
76927-132EA 290.69 USD
76927-132 76927-134 76927-136 76927-138
5-Hydroxy-2-methoxy-4-methylpyridine ≥97%
5-Hydroxy-2-methoxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11046206
CAS Number: 1086389-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR