(4-Chloro-2-fluoro-3-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-fluoro-3-methylphenylboronic acid

20471-1G 20471-25G 20471-5G
76927-112EA 106.09 USD
76927-112 76927-114 76927-116
(4-Chloro-2-fluoro-3-methylphenyl)boronic acid ≥97% research grade
(4-Chloro-2-fluoro-3-methylphenyl)boronic acid
Formula: C₇H₇BClFO₂
Storage Temperature: Ambient
MDL Number: MFCD18447609
CAS Number: 944128-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR