4-Ethoxy-2-fluorobenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

20432-1G 20432-25G 20432-5G
76927-084EA 62.6 USD
76927-084 76927-086 76927-088
4-Ethoxy-2-fluorobenzaldehyde ≥97% research grade
4-Ethoxy-2-fluorobenzaldehyde
Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06247347
CAS Number: 532965-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR