(4-Butoxy-2-fluorophenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-N-Butoxy-2-fluorophenylboronic acid

20295-25G 20295-5G
76927-068EA 595.17 USD
76927-068 76927-070
(4-Butoxy-2-fluorophenyl)boronic acid ≥97% research grade
(4-Butoxy-2-fluorophenyl)boronic acid
Formula: C₁₀H₁₄BFO₃
MW: 212.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08701767
CAS Number: 1107603-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR