Dibenzo[b,d]thiophene-4-carboxylic acid ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-Dibenzothiophenecarboxylic acid
20256-25G
20256-5G
76927-060EA
233.4
USD
76927-060
76927-062
Dibenzo[b,d]thiophene-4-carboxylic acid ≥97% research grade
Dibenzo[b,d]thiophene-4-carboxylic acid
Formula:
C₁₃H₈O₂S Storage Temperature: Ambient |
MDL Number:
MFCD00092754 CAS Number: 2786-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...