5-Chloro-2-fluoro-3-iodopyridine ≥97% research grade

Supplier: AOBChem USA

Warning

20189-1G 20189-25G 20189-500MG 20189-5G
76927-038EA 224.92 USD
76927-038 76927-040 76927-042 76927-044
5-Chloro-2-fluoro-3-iodopyridine ≥97% research grade
5-Chloro-2-fluoro-3-iodopyridine
Formula: C₅H₂ClFIN
Storage Temperature: Refrigerator
MDL Number: MFCD08277269
CAS Number: 884495-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR