4-Bromo-3-chlorophenylboronic acid ≥96%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromo-3-chlorophenyl)boronic acid

20142-1G 20142-25G 20142-5G
76927-020EA 62.6 USD
76927-020 76927-022 76927-024
4-Bromo-3-chlorophenylboronic acid ≥96%
4-Bromo-3-chlorophenylboronic acid
Formula: C₆H₅BBrClO₂
MW: 235.2700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12913945
CAS Number: 1217501-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR