5-Fluoro-2-(methylthio)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20135-1G 20135-25G 20135-500MG 20135-5G
76927-004EA 132.62 USD
76927-004 76927-006 76927-008 76927-010
5-Fluoro-2-(methylthio)phenylboronic acid ≥97%
5-Fluoro-2-(methylthio)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₂S
Storage Temperature: Ambient
MDL Number: MFCD15143469
CAS Number: 1218790-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR