4-Bromo-3-carboxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20100-1G 20100-25G 20100-5G
76926-994EA 106.09 USD
76926-994 76926-996 76926-998
4-Bromo-3-carboxyphenylboronic acid ≥97%
4-Bromo-3-carboxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BBrO₄
MW: 244.835 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17011990
CAS Number: 1451393-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR