5-Bromo-2-ethoxynicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

20055-1G 20055-25G 20055-500MG 20055-5G
76926-978EA 264.17 USD
76926-978 76926-980 76926-982 76926-984
5-Bromo-2-ethoxynicotinaldehyde ≥97%
5-Bromo-2-ethoxynicotinaldehyde

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD17011988
CAS Number: 1242070-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR