5-Bromo-2-fluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

20038-1G 20038-25G 20038-5G
76926-964EA 62.6 USD
76926-964 76926-966 76926-968
5-Bromo-2-fluoro-4-methylbenzaldehyde ≥97%
5-Bromo-2-fluoro-4-methylbenzaldehyde
Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD04112582
CAS Number: 497224-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR