5-Bromo-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-2-methylphenyl)boronic acid

19993-1G 19993-25G 19993-500MG 19993-5G
76926-948EA 290.69 USD
76926-948 76926-950 76926-952 76926-954
5-Bromo-2-methylphenylboronic acid ≥97%
5-Bromo-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD16295061
CAS Number: 774608-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR