5-Benzyloxy-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

19941-1G 19941-25G 19941-5G
76926-924EA 82.76 USD
76926-924 76926-926 76926-928
5-Benzyloxy-2-fluorophenylboronic acid ≥97%
5-Benzyloxy-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD13195645
CAS Number: 1217500-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR