5-Bromo-1,3-dichloro-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

19897-25G 19897-5G
76926-916EA 488.02 USD
76926-916 76926-918
5-Bromo-1,3-dichloro-2-isopropoxybenzene ≥97%
5-Bromo-1,3-dichloro-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD11855953
CAS Number: 1133116-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR